MilliporeSigma
  • Highly selective nickel-catalyzed three-component coupling of alkynes with enones and alkenyl boronic acids: a novel route to substituted 1,3-dienes.
Highly selective nickel-catalyzed three-component coupling of alkynes with enones and alkenyl boronic acids: a novel route to substituted 1,3-dienes.
Organic letters (2010-08-14)
Chun-Ming Yang, Masilamani Jeganmohan, Kanniyappan Parthasarathy, Chien-Hong Cheng
ABSTRACT
A highly regio- and stereoselective nickel-catalyzed three-component coupling of alkynes with enones and alkenyl boronic acids to afford highly substituted 1,3-dienes is described. The reaction can also be extended to cyclization of enynes with coupling to alkenyl boronic acids. A possible reaction mechanism involving a five-membered nickelacycle as a key intermediate is proposed.
Sigma-Aldrich
trans-1-Propen-1-ylboronic acid, ≥95.0%