MilliporeSigma
  • Emerging issues in receptor protein tyrosine phosphatase function: lifting fog or simply shifting?
Emerging issues in receptor protein tyrosine phosphatase function: lifting fog or simply shifting?
Journal of cell science (2000-06-15)
A Petrone, J Sap
ABSTRACT
Transmembrane (receptor) tyrosine phosphatases are intimately involved in responses to cell-cell and cell-matrix contact. Several important issues regarding the targets and regulation of this protein family are now emerging. For example, these phosphatases exhibit complex interactions with signaling pathways involving SRC family kinases, which result from their ability to control phosphorylation of both activating and inhibitory sites in these kinases and possibly also their substrates. Similarly, integrin signaling illustrates how phosphorylation of a single protein, or the activity of a pathway, can be controlled by multiple tyrosine phosphatases, attesting to the intricate integration of these enzymes in cellular regulation. Lastly, we are starting to appreciate the roles of intracellular topology, tyrosine phosphorylation and oligomerization among the many mechanisms regulating tyrosine phosphatase activity.