MilliporeSigma
  • Self-assembled monolayer of graphene/Pt as counter electrode for efficient dye-sensitized solar cell.
Self-assembled monolayer of graphene/Pt as counter electrode for efficient dye-sensitized solar cell.
Physical chemistry chemical physics : PCCP (2011-09-13)
Feng Gong, Hong Wang, Zhong-Sheng Wang
ABSTRACT
Monolayer of PDDA/graphene/PDDA/H(2)PtCl(6) is fabricated on conductive glass using electrostatic layer-by-layer self-assembly technique, which is then converted to graphene/Pt monolayer for use as counter electrode in dye-sensitized solar cell (DSSC). As compared to the sputtered Pt counter electrode, the self-assembled monolayer reduces the Pt amount by about 1000-fold but exhibits comparable photovoltaic performance. This finding provides a new route to fabrication of cheap and efficient counter electrodes for flow-line production of DSSCs.
Sigma-Aldrich
Platinum black, low bulk density, ≥99.9% trace metals basis
Sigma-Aldrich
Platinum on alumina, extent of labeling: 0.5 wt. % loading, spheres