MilliporeSigma
  • Hepatitis C virus-encoded enzymatic activities and conserved RNA elements in the 3' nontranslated region are essential for virus replication in vivo.
Hepatitis C virus-encoded enzymatic activities and conserved RNA elements in the 3' nontranslated region are essential for virus replication in vivo.
Journal of virology (2000-01-22)
A A Kolykhalov, K Mihalik, S M Feinstone, C M Rice
ABSTRACT
Hepatitis C virus (HCV) infection is a widespread major human health concern. Significant obstacles in the study of this virus include the absence of a reliable tissue culture system and a small-animal model. Recently, we constructed full-length HCV cDNA clones and successfully initiated HCV infection in two chimpanzees by intrahepatic injection of in vitro-transcribed RNA (A. A. Kolykhalov et al., Science 277:570-574, 1997). In order to validate potential targets for development of anti-HCV therapeutics, we constructed six mutant derivatives of this prototype infectious clone. Four clones contained point mutations ablating the activity of the NS2-3 protease, the NS3-4A serine protease, the NS3 NTPase/helicase, and the NS5B polymerase. Two additional clones contained deletions encompassing all or part of the highly conserved 98-base sequence at the 3' terminus of the HCV genome RNA. The RNA transcript from each of the six clones was injected intrahepatically into a chimpanzee. No signs of HCV infection were detected in the 8 months following the injection. Inoculation of the same animal with nonmutant RNA transcripts resulted in productive HCV infection, as evidenced by viremia, elevated serum alanine aminotransferase, and HCV-specific seroconversion. These data suggest that these four HCV-encoded enzymatic activities and the conserved 3' terminal RNA element are essential for productive replication in vivo.
Sigma-Aldrich
HCV-NS4A/NS3-1b Protease, Histag, strain HC-J4 from hepatitis C virus, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)
Sigma-Aldrich
HCV-NS4A/NS3-1a Protease, strain H77 from hepatitis C virus, recombinant, expressed in E. coli, ≥80% (SDS-PAGE)