MilliporeSigma
  • Microenvironment-Mediated Mechanisms of Resistance to HER2 Inhibitors Differ between HER2+ Breast Cancer Subtypes.
Microenvironment-Mediated Mechanisms of Resistance to HER2 Inhibitors Differ between HER2+ Breast Cancer Subtypes.
Cell systems (2018-03-20)
Spencer S Watson, Mark Dane, Koei Chin, Zuzana Tatarova, Moqing Liu, Tiera Liby, Wallace Thompson, Rebecca Smith, Michel Nederlof, Elmar Bucher, David Kilburn, Matthew Whitman, Damir Sudar, Gordon B Mills, Laura M Heiser, Oliver Jonas, Joe W Gray, James E Korkola
ABSTRACT
Extrinsic signals are implicated in breast cancer resistance to HER2-targeted tyrosine kinase inhibitors (TKIs). To examine how microenvironmental signals influence resistance, we monitored TKI-treated breast cancer cell lines grown on microenvironment microarrays composed of printed extracellular matrix proteins supplemented with soluble proteins. We tested ∼2,500 combinations of 56 soluble and 46 matrix microenvironmental proteins on basal-like HER2+ (HER2E) or luminal-like HER2+ (L-HER2+) cells treated with the TKIs lapatinib or neratinib. In HER2E cells, hepatocyte growth factor, a ligand for MET, induced resistance that could be reversed with crizotinib, an inhibitor of MET. In L-HER2+ cells, neuregulin1-β1 (NRG1β), a ligand for HER3, induced resistance that could be reversed with pertuzumab, an inhibitor of HER2-HER3 heterodimerization. The subtype-specific responses were also observed in 3D cultures and murine xenografts. These results, along with bioinformatic pathway analysis and siRNA knockdown experiments, suggest different mechanisms of resistance specific to each HER2+ subtype: MET signaling for HER2E and HER2-HER3 heterodimerization for L-HER2+ cells.
Sigma-Aldrich
Lapatinib
Sigma-Aldrich
MISSION® esiRNA, targeting human ERBB3
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Rabbit