MilliporeSigma
  • Effect of a mouthrinse containing Malva sylvestris on the viability and activity of microcosm biofilm and on enamel demineralization compared to known antimicrobials mouthrinses.
Effect of a mouthrinse containing Malva sylvestris on the viability and activity of microcosm biofilm and on enamel demineralization compared to known antimicrobials mouthrinses.
Biofouling (2018-02-13)
Aline Silva Braga, Juliana Gonçalves Pires, Ana Carolina Magalhães
ABSTRACT
The purpose of this study was to evaluate the antimicrobial (anti-biofilm) and anti-caries (enamel demineralization prevention) effects of Malva sylvestris (Malvatricin® Plus) compared with known antimicrobial mouthrinses. Microcosm biofilm was produced on enamel, using inoculum from pooled human saliva mixed with McBain saliva (0.2% sucrose) for 14 days. The biofilm was treated with mouthrinses for 1 min day-1. Oral-B® Complete, Listerine® Zero and Malvatricin® Plus had the greatest effect on the reduction of biofilm viability (p < 0.0001). On the other hand, lactic acid production was reduced significantly with PerioGard®, Noplak® Max and Listerine® Zero compared with the control (p < 0.0001). No significant differences were found among the mouthrinses with respect to the colony-forming unit counting (total microorganisms, total streptococci, mutans streptococci and lactobacilli) and extracellular polysaccharide production. Enamel demineralization was reduced significantly with PerioGard®, Noplak® Max and Malvatricin® Plus compared with the control (p < 0.0001). Malva sylvestris has a comparable anti-caries effect to chlorhexidine mouthrinses.
Sigma-Aldrich
Sodium fluorophosphate, 95%