MilliporeSigma
  • Amnionless-mediated glycosylation is crucial for cell surface targeting of cubilin in renal and intestinal cells.
Amnionless-mediated glycosylation is crucial for cell surface targeting of cubilin in renal and intestinal cells.
Scientific reports (2018-02-07)
Tomohiro Udagawa, Yutaka Harita, Kenichiro Miura, Jun Mitsui, Koji L Ode, Shinichi Morishita, Seiya Urae, Shoichiro Kanda, Yuko Kajiho, Haruko Tsurumi, Hiroki R Ueda, Shoji Tsuji, Akihiko Saito, Akira Oka
ABSTRACT
Mutations in either cubilin (CUBN) or amnionless (AMN) genes cause Imerslund-Gräsbeck syndrome (IGS), a hereditary disease characterised by anaemia attributed to selective intestinal malabsorption of cobalamin and low-molecular weight proteinuria. Although cubilin protein does not have a transmembrane segment, it functions as a multi-ligand receptor by binding to the transmembrane protein, amnionless. We established a system to quantitatively analyse membrane targeting of the protein complex in cultured renal and intestinal cells and analysed the pathogenic mechanisms of mutations found in IGS patients. A novel CUBN mutation, several previously reported CUBN missense mutations and all previously reported AMN missense mutations resulted in endoplasmic reticulum (ER) retention and completely inhibited amnionless-dependent plasma membrane expression of cubilin. The ER retention of cubilin and amnionless was confirmed in renal proximal tubular cells of a patient with IGS. Notably, the interaction between cubilin and amnionless was not sufficient, but amnionless-mediated glycosylation of cubilin was necessary for their surface expression. Quantitative mass spectrometry and mutagenesis demonstrated that N-linked glycosylation of at least 4 residues of cubilin protein was required for its surface targeting. These results delineated the molecular mechanisms of membrane trafficking of cubilin in renal and intestinal cells.
Sigma-Aldrich
Anti-AMN antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution