Skip to Content
MilliporeSigma
Methyl 3,4-O-isopropylidene-L-threonate 97%

Methyl 3,4-O-isopropylidene-L-threonate

Methyl 3,4-O-isopropylidene-L-threonate 97%
Synonym(s):
R,4S)-2,2-Dimethyl-α-hydroxy-1,3-dioxolane-4-acetic acid methyl ester, 3,4-O-Isopropylidene-L-threonic acid methyl ester
Empirical Formula (Hill Notation):
C8H14O5
CAS Number:
Molecular Weight:
190.19

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing