Skip to Content
MilliporeSigma
Isovaleraldehyde analytical standard

Isovaleraldehyde

Isovaleraldehyde analytical standard
Synonym(s):
3-Methylbutanal, 3-Methylbutyraldehyde, NSC 404119
Linear Formula:
(CH3)2CHCH2CHO
CAS Number:
Molecular Weight:
86.13
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing