
Eupalitin

Synonym(s):
3,4′,5-Trihydroxy-6,7-dimethoxyflavone, 3,5-Dihydroxy-2-(4-hydroxyphenyl)-6,7-dimethoxy-4H-1-benzopyran-4-one, 3,5-Dihydroxy-2-(4-hydroxyphenyl)-6,7-dimethoxychromen-4-one, Betuletol, Eupaletin
Empirical Formula (Hill Notation):
C17H14O7
CAS Number:
Molecular Weight:
330.29
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing