Skip to Content
MilliporeSigma
Cholic acid-24-13C 99 atom % 13C, 98% (CP), endotoxin tested

Cholic acid-24-13C

Cholic acid-24-13C 99 atom % 13C, 98% (CP), endotoxin tested
Empirical Formula (Hill Notation):
13CC23H40O5
CAS Number:
Molecular Weight:
409.56

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Technique
Showing 1-2 of 2