Key Documents
GBL103260
Grace Bio-Labs CultureWell adapter tray
holds 4 x 16 well chamber slides
Synonym(s):
chambered coverglass adapter
About This Item
UNSPSC Code:
41122600
NACRES:
NB.22
material
black polystyrene
sterility
non-sterile
packaging
pack of 2 ea
manufacturer/tradename
Grace Bio-Labs 103260
external L × W
127.5 mm × 85.5 mm
General description
The adapter tray fits most universal microscope adapters and is compatible with high content screening instruments.
CultureWell Microplate Adapter Tray holds up to 4 CultureWell 16 Chambers for higher throughput.
Legal Information
Culturewell is a trademark of Grace Bio-Labs, Inc.