Skip to Content
MilliporeSigma
All Photos(1)

Documents

Z268135

Sigma-Aldrich

Aldrich® desiccator cabinet

H × W × D 7 in. × 7 in. × 10 in., 2 trays, clear

Synonym(s):

Drying cabinet

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104510
NACRES:
NB.43

H × W × D

7 in. × 7 in. × 10 in.

color

clear

Looking for similar products? Visit Product Comparison Guide

General description

Aldrich desiccator cabinets are dust and moisture-free storage for refrigerators, freezers, and at room temperature. The clear walls, top, and base are 1/4 in. thick in acrylic for 360° visability.

Gasketed, air-tight doors are 3/8 in. thick acrylic with full length hinges and SS twist locking latches. Clear acrylic, deep slide-in trays with perforated bottoms for proper circulation within the cabinet. Dark bronze acrylic provides maximum protection for light-sensitive materials.

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service