Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

916749

Sigma-Aldrich

Pistons

3 mL, white, 100/box

Synonym(s):

3d bioprinting consumables

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26101736
NACRES:
NA.23

Quality Level

description

3mL pistons (100/box), white
Suitable for 3mL cartridge

Application

These are wiper pistons compatible with most fluids. A piston is inserted into the cartridge after it has been loaded with fluid to ensure uniform dispensing, prevent dripping, and eliminate waste by wiping barrel walls clean as fluid is dispensed.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service