- Cortactin is a sensitive biomarker relative to the poor prognosis of human hepatocellular carcinoma.
Cortactin is a sensitive biomarker relative to the poor prognosis of human hepatocellular carcinoma.
Cortactin is an important regulator involved in invasion and migration of hepatocellular carcinoma (HCC). The aim of this study was to elucidate the forecasting role of cortactin in resectable HCCs. We compared the invasiveness and motility among liver epithelial cell line and HCC cell lines by using Transwell assay and wound healing assay. We further investigated the CTTN mRNA expression by real-time PCR. Next, 91 HCC and 20 normal liver tissue samples were detected by IHC and real-time PCR. Finally, we analyzed the clinicopathologic features and survival time of the HCC cases. We identified that HepG2, LM3, and SK-Hep-1 had more invasiveness and motility (P <0.05). Compared with liver epithelial cell line, CTTN expression was higher in LM3, HepG2, and MHCC97-L (P <0.01) and lower in SK-Hep-1 (P <0.05). IHC examination showed cortactin expression was closely relative to TNM stage (AJCC/UICC), cancer embolus, and metastasis (P <0.01). Cortactin overexpression indicated a longer survival time of 52 ± 8.62 months and low expression of a shorter survival time of 20 ± 4.95 months (P <0.01). Cortactin examination has more predictive power in patients with Child-Pugh grade A and BCLC stage 0-B. Overexpression of cortactin is closely associated with poor human HCCs prognosis that caused by cancer embolus and metastasis. Cortactin and CTTN should be used for differentiating varieties of survival for patients after HCC resection.