Merck
  • Bioactivity-guided separation of anti-acidogenic substances against Streptococcus mutans UA 159 from Polygonum cuspidatum.
Bioactivity-guided separation of anti-acidogenic substances against Streptococcus mutans UA 159 from Polygonum cuspidatum.
Oral diseases (2010-04-09)
Y-R Kwon, K-J Son, S Pandit, J-E Kim, K-W Chang, J-G Jeon
ABSTRACT
The aim of this study was to separate the anti-acidogenic substances against Streptococcus mutans UA 159 from Polygonum cuspidatum. The anti-acidogenic substances were separated by a series of liquid-liquid fractionations followed by normal-phase silica gel liquid chromatography, based on high-performance liquid chromatography and glycolytic pH-drop assay. The effectiveness of the separated substances on the acidogenicity of Streptococcus mutans UA 159 was examined using sodium fluoride as a positive control. The chemical composition and quantities of the components of the substances was also assessed by qualitative-quantitative chromatographic analysis. Among the substances separated from P. cuspidatum, F3 showed the strongest inhibitory effect on the acidogenicity of S. mutans UA 159 in a dose-dependent manner without displaying any bactericidal activity. F3 decreased the acidogenicity of S. mutans even at 12.5 microg ml(-1) (P < 0.05). F3 consisted mainly of resveratrol and emodin (C(14)H(12)O(3) and C(14)H(4)O(2)(OH)(3)CH(3), respectively), which made up approximately 60% of the weight of F3. F3 can be considered as a promising agent for controlling the acidogenicity of S. mutans and subsequent dental caries formation.
Sigma-Aldrich
Physcion, ≥98.0% (TLC)
Supelco
Physcion, analytical standard