
Withanone

Synonym(s):
(22R)-6α,7α-Epoxy-5,17,22-trihydroxy-1-oxo-5α-ergosta-2,24-dien-26-oic acid δ-lactone
Empirical Formula (Hill Notation):
C28H38O6
CAS Number:
Molecular Weight:
470.60
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing