
Sabouraud Dextrose broth

Products
Product Category
Brand
Color
Physical Form
Agency
Chemical Composition
Capacity (mL)
Quality Segment
Closure Type
Application
Microorganism Suitability
Package Size
Sterility
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing