
Meclozine hydrochloride

Synonym(s):
Meclizine dihydrochloride, 1-([4-Chlorophenyl]phenylmethyl)-4-([3-methylphenyl]methyl)piperazine
Empirical Formula (Hill Notation):
C25H27ClN2 · 2HCl
CAS Number:
Molecular Weight:
463.87
Products
Formula Weight
Gene Alias
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing