Products
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Chemical Composition
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing