
Indocyanine green

Synonym(s):
Cardiogreen, Foxgreen, IC Green, Indocyanine green
Empirical Formula (Hill Notation):
C43H47N2NaO6S2
CAS Number:
Molecular Weight:
774.96
Products
Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing