Skip to Content
Merck
4-Nitrophenyl phosphate di(tris) salt phosphatase substrate

4-Nitrophenyl phosphate di(tris) salt

4-Nitrophenyl phosphate di(tris) salt phosphatase substrate
Synonym(s):
4-Nitrophenyl phosphate bis(tris) salt
Linear Formula:
(C6 H4 N O6 P)( C4 H12 N O3)2
CAS Number:
Molecular Weight:
461.36
EC Number:

Products

Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1