Merck
Ion-channel aligned gas-blocking membrane for lithium-air batteries.
Scientific reports (2017-09-22)
Wonsung Choi, Mokwon Kim, Jung Ock Park, Joon-Hee Kim, Kyunghwan Choi, Yong Su Kim, Tae Young Kim, Ken Ogata, Dongmin Im, Seok-Gwang Doo, Yunil Hwang
RESUMEN
Lithium-metal-based batteries, owing to the extremely high specific energy, have been attracting intense interests as post-Li-ion batteries. However, their main drawback is that consumption/de-activation of lithium metal can be accelerated when O
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
1,3,5-Triallyl-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, 98%
Sigma-Aldrich
2,2′:5′,2′′:5′′,2′′′-Quaterthiophene, 96%