Merck
Carrier-mediated transport controls hydroxyproline catabolism in heart mitochondria from spontaneously hypertensive rat.
FEBS letters (1996-11-04)
A Atlante, T M Seccia, E Marra, G M Minervini, V Vulpis, A Pirrelli, S Passarella
RESUMEN
In this study we have investigated hydroxyproline transport in rat heart mitochondria and, in particular, in heart left ventricle mitochondria isolated from both spontaneously hypertensive and Wistar-Kyoto rats. Hydroxyproline uptake by mitochondria, where its catabolism takes place, occurs via a carrier-mediated process as demonstrated by the occurrence of both saturation kinetics and the inhibition shown by phenylsuccinate and the thiol reagent mersalyl. In any case, hydroxyproline transport was found to limit the rate of mitochondrial hydroxyproline catabolism. A significant change in Vmax and Km values was found in mitochondria from hypertensive/hypertrophied rats in which the Km value decreases and the Vmax value increases with respect to normotensive rats, thus accounting for the increase of hydroxyproline metabolism due to its increased concentration in a hypertrophic/hypertensive state.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Phenylsuccinic acid, 98%