Merck
  • Determination of 3-chloropropane-1,2-diol in liquid hydrolyzed vegetable proteins and soy sauce by solid-phase microextraction and gas chromatography/mass spectrometry.
Determination of 3-chloropropane-1,2-diol in liquid hydrolyzed vegetable proteins and soy sauce by solid-phase microextraction and gas chromatography/mass spectrometry.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry (2005-12-02)
Minjia Huang, Guibin Jiang, Bin He, Jingfu Liu, Qingxiang Zhou, Wusheng Fu, Yongning Wu
RESUMEN
Headspace solid-phase microextraction (HS-SPME) coupled with gas chromatography/mass spectrometry (GC/MS) method was developed to determine 3-chloropropane-1,2-diol (3-MCPD) in hydrolyzed vegetable protein and Chinese soy sauce. The 3-MCPD was firstly derivativized with phenylboronic acid in aqueous solution at 90 degrees C for 10 min, then extracted by HS-SPME and finally detected with GC/MS, parameters related to both the derivative reaction and the HS-SPME process were optimized. The proposed method has a linear range of 0.0194-394 microg g(-1), a detection limit of 3.87 ng g(-1) (S/N = 3), and a precision of RSD = 7.5% (n = 5). Seventeen real samples, including four HVPs and thirteen soy sauce samples, were analyzed to examine the feasibility of the proposed procedure; with a concentration of 3-MCPD and acceptable recoveries at 0.71 microg g(-1) spiked levels were obtained. Being simpler, faster and more environmentally benign than the existing methods, this method is accurate and suitable for routine analysis.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
N-Heptafluorobutyrylimidazole, 97%
Sigma-Aldrich
1-(Heptafluorobutyryl)imidazole, BioReagent, suitable for derivatization