Merck
  • Validation of method for determination of different classes of pesticides in aqueous samples by dispersive liquid-liquid microextraction with liquid chromatography-tandem mass spectrometric detection.
Validation of method for determination of different classes of pesticides in aqueous samples by dispersive liquid-liquid microextraction with liquid chromatography-tandem mass spectrometric detection.
Analytica chimica acta (2010-04-13)
Sergiane Souza Caldas, Fabiane Pinho Costa, Ednei Gilberto Primel
RESUMEN
In this study, a simple, rapid and efficient method has been developed for the extraction and preconcentration of different classes of pesticides, carbofuran (insecticide), clomazone (herbicide) and tebuconazole (fungicide) in aqueous samples by dispersive liquid-liquid microextraction (DLLME) coupled with liquid chromatography-tandem mass spectrometric detection. Some experimental parameters that influence the extraction efficiency, such as the type and volume of the disperser solvents and extraction solvents, extraction time, speed of centrifugation, pH and addition of salt were examined and optimized. Under the optimum conditions, the recoveries of pesticides in water at spiking levels between 0.02 and 2.0 microg L(-1) ranged from 62.7% to 120.0%. The relative standard deviations varied between 1.9% and 9.1% (n=3). The limits of quantification of the method considering a 50-fold preconcentration step were 0.02 microg L(-1). The linearity of the method ranged from 1.0 to 1000 microg L(-1) for all compounds, with correlation coefficients varying from 0.9982 to 0.9992. Results show that the method we propose can meet the requirements for the determination of pesticides in water samples. The comparison of this method with solid-phase extraction indicates that DLLME is a simple, fast, and low-cost method for the determination of pesticides in natural waters.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Supelco
Clomazone, PESTANAL®, analytical standard