Merck
3-Chloro-DL-alanine resistance by L-methionine-alpha-deamino-gamma-mercaptomethane-lyase activity.
FEBS letters (2002-07-19)
Mamiko Yoshimura, Yoshio Nakano, Haruka Fukamachi, Toshihiko Koga
RESUMEN
The antibacterial agent 3-chloro-DL-alanine (3CA) is an inhibitor of peptidoglycan synthesis. Fusobacterium nucleatum and Porphyromonas gingivalis, the bacteria responsible for oral malodor, are shown to be resistant to 1 mM 3CA, whereas Streptococcus mutans and Escherichia coli are sensitive to this antibacterial agent at the same concentration. We isolated the 3CA resistance gene from F. nucleatum and showed that the gene encodes an L-methionine-alpha-deamino-gamma-mercaptomethane-lyase that catalyzes the alpha,gamma-elimination of L-methionine to produce methyl mercaptan. The enzyme also exhibits 3CA chloride-lyase (deaminating) activity. This antibacterial agent is expected to be useful for specific selection of malodorous oral bacteria producing high amounts of methyl mercaptan.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
β-Chloro-L-alanine hydrochloride, Alanine aminotransferase inhibitor
Sigma-Aldrich
β-Chloro-D-alanine hydrochloride