Merck
Self-assembly of a metal-ion-bound monolayer of trigonal connectors on mercury: an electrochemical Langmuir trough.
Proceedings of the National Academy of Sciences of the United States of America (2002-04-18)
Natalia Varaksa, Lubomír Pospísil, Thomas F Magnera, Josef Michl
RESUMEN
The adsorption of the trigonal connector, 1,3,5-tris[10-(3-ethylthiopropyl)dimethylsilyl-1,10-dicarba-closo-decaboran-1-yl]benzene (1), from acetonitrile/0.1 M LiClO(4) on the surface of mercury at potentials ranging from +0.3 to -1.4 V (vs. aqueous Ag/AgCl/1 M LiCl) was examined by voltammetry, Langmuir isotherms at controlled potentials, and impedance measurements. No adsorption is observed at potentials more negative than approximately -0.85 V. Physisorption is seen between approximately -0.85 and 0 V. At positive potentials, adsorbate-assisted anodic dissolution of mercury occurs and an organized surface layer is formed. Although the mercury cations are reduced at -0.10 V, the surface layer remains metastable to potentials as negative as -0.85 V. Its surface areas per molecule and per redox center are compatible with a regular structure with the connectors 1 woven into a hexagonal network by RR'S-->Hg(2)(2+)<--SRR' or RR'S-->Hg(2+)<--SRR' bridges. The structure is simulated closely by geometry optimization in the semiempirical AM1 approximation.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Lithium perchlorate, ACS reagent, ≥95.0%
Sigma-Aldrich
Lithium perchlorate, battery grade, dry, 99.99% trace metals basis
Sigma-Aldrich
Lithium perchlorate, 99.99% trace metals basis
Sigma-Aldrich
Lithium perchlorate, purum p.a., ≥98.0% (calc. based on dry substance, T), powder
Sigma-Aldrich
Lithium perchlorate, SAJ special grade, ≥99.0%