Merck
Nanoparticle-Mediated Recombinase Delivery into Maize.
Methods in molecular biology (Clifton, N.J.) (2017-08-18)
Susana Martin-Ortigosa, Brian G Trewyn, Kan Wang
RESUMEN
We describe a non-DNA-based system for delivering Cre recombinase protein into maize tissue using gold-plated mesoporous silica nanoparticle (Au-MSN). Cre protein is first loaded into the pores of Au-MSNs and then delivered using the biolistic method to immature embryos of a maize line (Lox-corn), which harbors loxP sites flanking a selection and a reporter gene. The release of the Cre recombinase protein inside the plant cell leads to recombination at the loxP sites, eliminating both genes. Visual screening is used to identify recombination events, which can be regenerated to mature and fertile plants. Using the experimental procedures and conditions described here, as high as 20% of bombarded embryos can produce regenerable recombinant callus events. This nanomaterial-mediated, DNA-free methodology has potential to become an effective tool for plant genome editing.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Anti-Cre antibody, Mouse monoclonal, clone 7-23, purified from hybridoma cell culture