Merck
A Combined Mechanochemical and Calcination Route to Mixed Cobalt Oxides for the Selective Catalytic Reduction of Nitrophenols.
Molecules (Basel, Switzerland) (2019-12-29)
Lorianne R Shultz, Bryan McCullough, Wesley J Newsome, Haider Ali, Thomas E Shaw, Kristopher O Davis, Fernando J Uribe-Romo, Matthieu Baudelet, Titel Jurca
RESUMEN
Para-, or 4-nitrophenol, and related nitroaromatics are broadly used compounds in industrial processes and as a result are among the most common anthropogenic pollutants in aqueous industrial effluent; this requires development of practical remediation strategies. Their catalytic reduction to the less toxic and synthetically desirable aminophenols is one strategy. However, to date, the majority of work focuses on catalysts based on precisely tailored, and often noble metal-based nanoparticles. The cost of such systems hampers practical, larger scale application. We report a facile route to bulk cobalt oxide-based materials, via a combined mechanochemical and calcination approach. Vibratory ball milling of CoCl2(H2O)6 with KOH, and subsequent calcination afforded three cobalt oxide-based materials with different combinations of CoO(OH), Co(OH)2, and Co3O4 with different crystallite domains/sizes and surface areas; Co@100, Co@350 and Co@600 (Co@###; # = calcination temp). All three prove active for the catalytic reduction of 4-nitrophenol and related aminonitrophenols. In the case of 4-nitrophenol, Co@350 proved to be the most active catalyst, therein its retention of activity over prolonged exposure to air, moisture, and reducing environments, and applicability in flow processes is demonstrated.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
4-Amino-3-nitrophenol, 98%