Merck
Apoptotic efficacy of multifaceted biosynthesized silver nanoparticles on human adenocarcinoma cells.
Scientific reports (2018-09-27)
Blassan Plackal Adimuriyil George, Neeraj Kumar, Heidi Abrahamse, Suprakas Sinha Ray
RESUMEN
Metallic nanoparticles (NPs) especially silver (Ag) NPs have shown immense potential in medical applications due to their distinctive physio-chemical and biological properties. This article reports the conjugation of Ag NPs with Rubus fairholmianus extract. The modification of Ag NPs was confirmed using various physico-chemical characterization techniques. The cytotoxic effect of Rubus-conjugated Ag NPs (RAgNPs) was studied by LDH assay and proliferation by ATP assay. The apoptotic inducing ability of the NPs were investigated by Annexin V/PI staining, caspase 3/7 analysis, cytochrome c release, intracellular ROS analysis, Hoechst staining and mitochondrial membrane potential analysis using flow cytometry. The expression of apoptotic proteins caspase 3, Bax and P53 were analyzed using ELISA and caspase 3, Bax using western blotting. Cells treated with 10 µg/mL RAgNPs showed an increased number of cell death by microscopic analysis compared to untreated control cells. The RAgNPs induced a statistically significant dose-dependent decrease in proliferation (p < 0.001 for 5 and 10 µg/mL) and increased cytotoxicity in MCF-7 cells. A 1.83 fold increase in cytotoxicity was observed in cells treated with 10 µg/mL (p < 0.05) compared to the untreated cells. Nuclear damage and intracellular ROS production were observed upon treatment with all tested concentrations of RAgNPs and the highest concentrations (5 and 10 µg/mL) showed significant (p < 0.05, p < 0.01) expression of caspase 3, Bax and P53 proteins. The data strongly suggest that RAgNPs induces cell death in MCF-7 cells through the mitochondrial-mediated intrinsic apoptosis pathway. The present investigation supports the potential of RAgNPs in anticancer drug development.
MATERIALES
Referencia del producto
Marca
Descripción del producto
Sigma-Aldrich
Propyl gallate, for microscopy, ≥98.0% (HPLC)
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad
Sigma-Aldrich
2-Phenylindole, technical grade, 95%
Número de referencia del producto (SKU)
Tamaño de envase
Disponibilidad
Precio
Cantidad