Merck
  • miR-9 regulates basal ganglia-dependent developmental vocal learning and adult vocal performance in songbirds.
miR-9 regulates basal ganglia-dependent developmental vocal learning and adult vocal performance in songbirds.
eLife (2018-01-19)
Zhimin Shi, Zoe Piccus, Xiaofang Zhang, Huidi Yang, Hannah Jarrell, Yan Ding, Zhaoqian Teng, Ofer Tchernichovski, XiaoChing Li
ABSTRACT
miR-9 is an evolutionarily conserved miRNA that is abundantly expressed in Area X, a basal ganglia nucleus required for vocal learning in songbirds. Here, we report that overexpression of miR-9 in Area X of juvenile zebra finches impairs developmental vocal learning, resulting in a song with syllable omission, reduced similarity to the tutor song, and altered acoustic features. miR-9 overexpression in juveniles also leads to more variable song performance in adulthood, and abolishes social context-dependent modulation of song variability. We further show that these behavioral deficits are accompanied by downregulation of
Sigma-Aldrich
Anti-FOXP2 antibody produced in rabbit, AB1, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Anti-FOXP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution