Merck
  • The influence of penicillamine/cysteine mutation on the metal complexes of peptides.
The influence of penicillamine/cysteine mutation on the metal complexes of peptides.
Dalton transactions (Cambridge, England : 2003) (2017-09-28)
Ágnes Grenács, Norbert Lihi, Imre Sóvágó, Katalin Várnagy
ABSTRACT
N-Terminally free but C-terminally amidated peptides Pen-SSACS-NH
Sigma-Aldrich
Fmoc-Pen(Trt)-OH, 96%
Sigma-Aldrich
Fmoc-Cys(Trt)-OH, ≥95.0% (sum of enantiomers, HPLC)