Merck
Partitioning and diffusion of parathion in human dermis.
International journal of pharmaceutics (2012-04-10)
Rania Ibrahim, Gerald B Kasting
ABSTRACT
A previously developed method employing the use of a dialysis membrane in series with human dermis tissue mounted in side-by-side diffusion cells was utilized to observe the effects of the presence of soluble proteins in the donor compartment on the measured transport parameters of parathion. In the presence of the dialysis membrane the partition coefficient was significantly lower and the diffusion coefficient significantly higher than those determined in its absence; however, the difference was less than that previously determined for the more highly protein bound compound, diclofenac. The result suggests the dialysis membrane method is important for studying permeants that are more than about 87% bound to soluble proteins in the dermis. The results are discussed in the context of a predictive model for partitioning and transport of low molecular weight solutes in human dermis.
Product Description
Supelco
Parathion solution, 100 μg/mL in cyclohexane, PESTANAL®, analytical standard
Supelco
Parathion, PESTANAL®, analytical standard