Merck
  • Convergent activation of two-pore channels mediated by the NAADP-binding proteins JPT2 and LSM12.
Convergent activation of two-pore channels mediated by the NAADP-binding proteins JPT2 and LSM12.
Science signaling (2023-08-22)
Gihan S Gunaratne, Eugen Brailoiu, Sushil Kumar, Yu Yuan, James T Slama, Timothy F Walseth, Sandip Patel, Jonathan S Marchant
ABSTRACT
The second messenger nicotinic acid adenine dinucleotide phosphate (NAADP) evokes calcium ion (Ca2+) release from endosomes and lysosomes by activating two-pore channels (TPCs) on these organelles. Rather than directly binding to TPCs, NAADP associates with proteins that indirectly confer NAADP sensitivity to the TPC complex. We investigated whether and how the NAADP-binding proteins Jupiter microtubule-associated homolog 2 (JPT2) and like-Sm protein 12 (LSM12) contributed to NAADP-TPC-Ca2+ signaling in human cells. Biochemical and functional analyses revealed that recombinant JPT2 and LSM12 both bound to NAADP with high affinity and that endogenous JPT2 and LSM12 independently associated with TPC1 and TPC2. On the basis of knockout and rescue analyses, both NAADP-binding proteins were required to support NAADP-evoked Ca2+ signaling and contributed to endolysosomal trafficking of pseudotyped coronavirus particles. These data reveal that the NAADP-binding proteins JPT2 and LSM12 convergently regulate NAADP-evoked Ca2+ release and function through TPCs.
Sigma-Aldrich
Anti-HN1L antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
25 g
Estimated to ship on March 23, 2025
MXP 2,865.00
100 g
Estimated to ship on March 23, 2025
MXP 8,909.00
500 g
Estimated to ship on March 23, 2025
MXP 19,563.00
To order products, please contact your local dealer.