Merck
Essential requirement for JPT2 in NAADP-evoked Ca2+ signaling.
Science signaling (2021-03-25)
Gihan S Gunaratne, Eugen Brailoiu, Shijun He, Ellen M Unterwald, Sandip Patel, James T Slama, Timothy F Walseth, Jonathan S Marchant
ABSTRACT
Nicotinic acid adenine dinucleotide phosphate (NAADP) is a second messenger that releases Ca2+ from acidic organelles through the activation of two-pore channels (TPCs) to regulate endolysosomal trafficking events. NAADP action is mediated by NAADP-binding protein(s) of unknown identity that confer NAADP sensitivity to TPCs. Here, we used a "clickable" NAADP-based photoprobe to isolate human NAADP-binding proteins and identified Jupiter microtubule-associated homolog 2 (JPT2) as a TPC accessory protein required for endogenous NAADP-evoked Ca2+ signaling. JPT2 was also required for the translocation of a severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) pseudovirus through the endolysosomal system. Thus, JPT2 is a component of the NAADP receptor complex that is essential for TPC-dependent Ca2+ signaling and control of coronaviral entry.
Sigma-Aldrich
Anti-HN1L antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPrice
25 g
Please contact Customer Service for Availability
CA$180.00
100 g
Please contact Customer Service for Availability
CA$600.00
To order products, please contact your local dealer.