
Tryptanthrine

Synonym(s):
Indolo(2,1-b)quinazoline-6,12-dione, Indolo[2,1-b]quinazoline-6,12-dione
Empirical Formula (Hill Notation):
C15H8N2O2
CAS Number:
Molecular Weight:
248.24
Products
Formula Weight
Manufacturer
Product Line
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing