
Bifonazole for system suitability

Synonym(s):
Bifonazole, 1-(p,α-Diphenylbenzyl)imidazole
Empirical Formula (Hill Notation):
C22H18N2
CAS Number:
Molecular Weight:
310.39
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing