
Anti-FOXP2 antibody produced in rabbit

Synonym(s):
Anti-CAG repeat protein 44 antibody produced in rabbit, Anti-Forkhead box protein P2 antibody produced in rabbit, Anti-Trinucleotide repeat-containing gene 10 protein antibody produced in rabbit
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Post Translational Modification
Species Reactivity
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing