Skip to Content
Merck
All Photos(1)

Key Documents

04-1651

Sigma-Aldrich

Rabbit Anti-Mouse Ig lambda light chain Antibody, clone RM110

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

Quality Level

clone

RM110
monoclonal

purified by

using Protein A

species reactivity

mouse, rabbit

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

General description

Mouse IgM Lambda

Specificity

This antibody reacts to the lamda light chain of mouse immunoglobulins. No cross reactivity with the kappa light chain, human IgG, rat IgG, or goat IgG. The Fc region of clone RM110 has been engineered to eliminate the binding of Fc receptor.

Application

ELISA Analysis: The specificity and reactivity of a representative lot was confirmed by ELISA analyses.

Quality

Evaluated by Western Blotting in nonreduced and reduced mouse IgG1κ and IgG1γ.

Western Blotting Analysis (WB): 0.2 μg/mL of this antibody detected Mouse Ig lambda light chain in nonreduced mouse IgG1γ.

Physical form

Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service