Merck
  • Solid phase extraction of chromium(VI) from aqueous solutions by adsorption of its diphenylcarbazide complex on an Amberlite XAD-4 resin column.
Solid phase extraction of chromium(VI) from aqueous solutions by adsorption of its diphenylcarbazide complex on an Amberlite XAD-4 resin column.
Journal of hazardous materials (2007-06-30)
N Rajesh, Rohit Kumar Jalan, Pinky Hotwany
ABSTRACT
A method has been developed for the solid phase extraction of chromium(VI) based on the adsorption of its diphenylcarbazide complex on an Amberlite XAD-4 resin column. The influence of acidity, stability of the column, sample volume, flow rate and interfering ions were studied in detail. The adsorbed complex could be eluted using acetone-sulfuric acid mixture and the concentration of chromium was determined using visible spectrophotometry. A detection limit of 6 microg L(-1) could be achieved. A preconcentration factor of 27 could be obtained for 400 mL sample volume. The validity of the method was checked in spiked water samples and electroplating wastewater.
Sigma-Aldrich
1,5-Diphenylcarbazide, JIS special grade
Sigma-Aldrich
1,5-Diphenylcarbazide, ACS reagent
Sigma-Aldrich
1,5-Diphenylcarbazide, reagent grade
Supelco
1,5-Diphenylcarbazide, reag. Ph. Eur., ≥98.0%