Merck
  • Genotoxic activation of benzophenone and its two metabolites by human cytochrome P450s in SOS/umu assay.
Genotoxic activation of benzophenone and its two metabolites by human cytochrome P450s in SOS/umu assay.
Mutation research (2002-08-06)
Kei Takemoto, Hiroshi Yamazaki, Miki Nakajima, Tsuyoshi Yokoi
ABSTRACT
The genotoxic potential of benzophenone and its metabolically related compounds, benzhydrol and p-benzoylphenol, was investigated using human cytochrome P450 (P450) enzymes. Benzophenone and its two metabolites (0.1-1mM) showed a suppression of bacterial growth without any P450 system, but no induction of umu gene expression was observed in Salmonella typhimurium TA1535/pSK1002. Human liver microsomes induced the bacterial cytotoxicity of these compounds without any umu gene expression. On the other hand, with the addition of Escherichia coli membranes expressing recombinant human P450 2A6 and NADPH-cytochrome P450 reductase (NPR), benzophenone showed umu gene expression (64 umu units/min/nmol) P450 2A6). Moderate activation of benzophenone by P450 1A1/NPR membranes, 1A2/NPR membranes, or 1B1/NPR membranes was also observed. Activation of benzhydrol and p-benzoylphenol by the P450/NPR system was similar to that of benzophenone. These results suggest that benzophenone and its metabolically related benzhydrol and p-benzoylphenol can be bioactivated by P450 2A6 and P450 family 1 enzymes. Until now, benzophenone has been investigated mainly in terms of estrogenic activity and cytotoxicity, however, the genotoxic activation of benzophenone by human cytochrome P450s should be examined in terms of the risks to humans.
Sigma-Aldrich
4-Benzylphenol, 99%
Pricing and availability is not currently available.
Sigma-Aldrich
Diphenylmethanol, 99%
Pricing and availability is not currently available.