
Maize GMO Standard MON 863 x MON 810

Synonym(s):
GMO Maize Powder Standard 0% MON 863 x MON 810
Products
Manufacturer
Product Line
Format
Chemical Composition
Application
Reference Material Type
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing