
(±)-Diclofop

Synonym(s):
(RS)-2-[4-(2,4-Dichlorophenoxy)phenoxy]propionic acid
Empirical Formula (Hill Notation):
C15H12Cl2O4
CAS Number:
Molecular Weight:
327.16
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2