
Cortisone-2,2,4,6,6,9,12,12-d8

Empirical Formula (Hill Notation):
C21D8H20O5
CAS Number:
Molecular Weight:
368.49
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Isotope
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing