
Chlorthal-dimethyl

Synonym(s):
Dimethyl tetrachloroterephthalate
Empirical Formula (Hill Notation):
C10H6Cl4O4
CAS Number:
Molecular Weight:
331.96
Products
Brand
Formula Weight
Manufacturer
Quality Segment
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing