
2-Methoxyestrone

Synonym(s):
2,3-Dihydroxy-1,3,5(10)-estratrien-17-one 2-methyl ether, 2-Hydroxyestrone 2-methyl ether, 3-Hydroxy-2-methoxy-1,3,5(10)-estratrien-17-one
Empirical Formula (Hill Notation):
C19H24O3
CAS Number:
Molecular Weight:
300.39
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing