Merck
  • Comparison of GIAO and CSGT for calculating (13) C and (15) N nuclear magnetic resonance chemical shifts of substituent neutral 5-aminotetrazole and 5-nitrotetrazole compounds.
Comparison of GIAO and CSGT for calculating (13) C and (15) N nuclear magnetic resonance chemical shifts of substituent neutral 5-aminotetrazole and 5-nitrotetrazole compounds.
Magnetic resonance in chemistry : MRC (2012-01-25)
Yubo Li, Haixiang Gao, Jiaheng Zhang, Songqing Li, Wenfeng Zhou
ABSTRACT
The comparison of the gauge-including atomic orbital (GIAO) and the continuous set of gauge transformation methods for calculating nuclear magnetic chemical shifts (CSs) mainly at density functional levels of theory are presented. Isotropic (13)  C and (15)  N magnetic CS for 14 compounds of tetrazoles are reported. Compared with establishing a convenient and consistent protocol to be employed for confirming the experimental (13)  C and (15)  N NMR spectra of tetrazole compounds, different combinations of functionals and basis sets were considered. The most reliable results were obtained at GIAO/B3LYP/aug-cc-pVDZ with integral equation formulation for the polarizable continuum model (PCM), which has the smallest root mean square errors and can be used to calculate (13)  C and (15)  N NMR CS with a very high accuracy for tetrazoles. These results show that the accurately calculated (15) N NMR CS of tetrazoles could be used for the evaluation of the intrinsic relationship between structure and explosive properties.
Sigma-Aldrich
5-Aminotetrazole, 97%