Merck
  • Crotonaldehyde induces apoptosis in alveolar macrophages through intracellular calcium, mitochondria and p53 signaling pathways.
Crotonaldehyde induces apoptosis in alveolar macrophages through intracellular calcium, mitochondria and p53 signaling pathways.
The Journal of toxicological sciences (2013-03-29)
Bi-cheng Yang, Xiu-jie Pan, Zhi-hua Yang, Feng-jun Xiao, Xing-yu Liu, Mao-xiang Zhu, Jian-ping Xie
ABSTRACT
Crotonaldehyde, a highly electrophilic α, β-unsaturated aldehyde, is a ubiquitous environmental pollutant and a risk factor for multiple respiratory diseases. Crotonaldehyde is highly volatile and hydrophilic, so it is efficiently absorbed in the respiratory tract. Alveolar macrophages are major effector cells of the nonspecific host defence in the lung. The aim of this study was to investigate the molecular mechanisms and signaling pathways responsible for cell death of alveolar macrophage induced by crotonaldehyde. Our results show that crotonaldehyde induces apoptosis in alveolar macrophages, as indicated by phosphatidylserine externalization and DNA fragmentation. Pretreatment of alveolar macrophages with N-acetylcysteine, ascorbic acid, α-tocopherol, superoxide dismutase inhibited crotonaldehyde-induced apoptosis. Crotonaldehyde-induced apoptosis was characterized by ROS generation, GSH depletion, loss of mitochondrial membrane potential (ΔΨm), the release of cytochrome c from mitochondria, caspase-3/7 and caspase-9 activation, elevation of intracellular Ca(2+) concentration and the increase of p53 expression. Furthermore, pretreatment with either p53 inhibitor pifithrin-α or calcium chelator BAPTA-AM effectively attenuated apoptosis induced by crotonaldehyde. Taken together, our results showed that crotonaldehyde induce apoptosis in alveolar macrophages through intracellular calcium, mitochondria and p53 signaling pathways. These results would help to illustrate the mechanism of toxicity induced by crotonaldehyde and to look for a novel treatment for diseases induced by exposure to crotonaldehyde-rich pollutants such as cigarette smoke.
Sigma-Aldrich
ProteoMass Cytochrome c MALDI-MS Standard, vial of 10 nmol, (M+H+) 12,361.96 Da by calculation
Sigma-Aldrich
Cytochrome c from Saccharomyces cerevisiae, ≥85% based on Mol. Wt. 12,588 basis
Sigma-Aldrich
Crotonaldehyde, mixture of cis and trans, ratio of cis- and trans-isomers (~1:20), ≥99.5% (GC)
Sigma-Aldrich
Crotonaldehyde, predominantly trans, ≥99%, contains 0.1-0.2% BHT as stabilizer, 1% H2O as stabilizer
Sigma-Aldrich
Cytochrome c from equine heart, BioUltra, ≥99% (SDS-PAGE), powder, suitable for mammalian cell culture
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% based on Mol. Wt. 12,384 basis
Sigma-Aldrich
Cytochrome c from equine heart, ≥95% (SDS-PAGE)
Sigma-Aldrich
Cytochrome c from equine heart, BioReagent, suitable for GFC marker
Sigma-Aldrich
Cytochrome c from bovine heart, ≥95% based on Mol. Wt. 12,327 basis, powder, suitable for mammalian cell culture
Sigma-Aldrich
Cytochrome c from pigeon breast muscle, ≥95% based on Mol. Wt. 12,173 basis