Merck
  • Detecting phthalate esters in sludge particulates from wastewater treatment plants.
Detecting phthalate esters in sludge particulates from wastewater treatment plants.
Journal of environmental science and health. Part A, Toxic/hazardous substances & environmental engineering (2020-06-20)
Cheng-Di Dong, Ming-Huang Wang, Chih-Feng Chen, Yu-Jen Shih, Ken-Lin Chang, Shu-Hui Lee, Yi-Li Lin, Chung-Hsin Wu, Chiu-Wen Chen
ABSTRACT
This study proposed a method for analysis of 10 phthalate esters compounds from wastewater treatment plant sludges. The analytical efficiency of GC-MS for of target compounds was verified by a standard mixture of phthalate esters. The response factors related to the respective internal standards from a five-point calibration curve quantified the phthalate esters in individual compounds. Based on the literature compiled by environmental agencies, new generation phthalate compounds have been developed, such as di-iso-nonyl phthalate (DiNP), di-iso-decyl phthalate (DiDP), as alternative to conventional phthalates. The analytical results showed that the total PAEs concentration was in the range from 7.4 to 138.6 mg kg-1 dw in these seven analyzed sludge samples. More, di-iso-nonyl phthalate (DiNP), di-iso-decyl phthalate (DiDP) and bis(2-ethylhexyl) phthalate (DEHP) contributed to over 99% of PAEs in the sludge. The correlation between total PAEs concentration in household and sewage flow treated at seven WWTPs and concentrations of DEHP, DiNP and DiDP was significant.
Sigma-Aldrich
Dess-Martin periodinane, 97%
Supelco
Dihexyl phthalate, analytical standard